Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MSQKPAKEG PRLSKNQKYS EHFSIHCCPP FTFLNSKKEI VDRKYSICKS GCFYQKKEED WICCACQKTR L (1-70 aa encoded by BC022471)
MOBP may play a role in compacting or stabilizing the myelin sheath, possibly by binding the negatively charged acidic phospholipids of the cytoplasmic membrane.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Myelin-associated oligodendrocyte basic protein; myelin-associated oligodendrocytic basic protein; Myelin-associated/Oligodendrocytic Basic Protein-81; RNMOBP81P
Gene Aliases: AI835053; AI839848; MOBP; MOBP155; MOBP170; MOBP69; MOBP73; MOBP81; Mobp81p; R74645
UniProt ID: (Human) Q6DHZ6, (Mouse) Q9D2P8, (Rat) Q63327
Entrez Gene ID: (Human) 4336, (Mouse) 17433, (Rat) 25037
Molecular Function: myelin protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support