Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MASLSRPSLP SCLCSFLLLL LLQVSSSYAG QFRVIGPRHP IRALVGDEVE LPCRISPGKN ATGMEVGWYR PPFSRVVHLY RNGKDQDGDQ APEYRGRTEL LKDAIGEGKV TLRIRNVRFS DEGGFTCFFR DHSYQEEAAM ELKVEDPFYW VSPGVLVLLA VLPVLLLQIT VGLVFLCLQY RLRGKLRAEI ENLHRTFDPH FLRVPCWKIT LFVIVPVLGP LVALIICYNW LHRRLAGQFL EELRKFSSLC YKQRIKSQER ETEATRGRGG LLRDHIPRGK EELESLGGGK TPPGR
The product of this gene is a membrane protein expressed on the oligodendrocyte cell surface and the outermost surface of myelin sheaths. Due to this localization, it is a primary target antigen involved in immune-mediated demyelination. This protein may be involved in completion and maintenance of the myelin sheath and in cell-cell communication. Alternatively spliced transcript variants encoding different isoforms have been identified.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: DAQB-92E24.2; MGC26137; MOG alpha 6; MOG alpha-5; MOG alpha-6; MOG AluA; MOG AluB; MOG Ig AluB; MOG Ig-AluB; MOGIG-2; Myelin-oligodendrocyte glycoprotein
Gene Aliases: BTN6; BTNL11; MOG; MOGIG2; NRCLP7
UniProt ID: (Human) Q5SUK7
Entrez Gene ID: (Human) 4340
Molecular Function:
immunoglobulin receptor superfamily
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support