Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 2
The recommended shelf life for this product is 1 year from date of receipt.
MON1B (MON1 Homolog B, Secretory Trafficking Associated) is a Protein Coding gene. Among its related pathways areVesicle-mediated transportandRAB GEFs exchange GTP for GDP on RABs. An important paralog of this gene isMON1A.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: HSV-1 stimulation-related gene 1 protein; HSV-I stimulating-related protein; MON1 homolog B; MON1 secretory trafficking family member B; Vacuolar fusion protein MON1 homolog B
Gene Aliases: HSRG1; KIAA0872; MON1B; SAND2; SRG1
UniProt ID: (Human) Q7L1V2
Entrez Gene ID: (Human) 22879
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support