Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MSSRKQGSQ PRGQQSAEEE NFKKPTRSNM QRSKMRGASS GKKTAGPQQK NLEPALPGRW GGRSAENPPS GSVRKTRKNK QKTPGNGDGG STSEAPQPPR KKRARADPTV ESEEAFKNRM EVKVKIPEEL KPWLVEDWDL VTRQKQLFQL PAKKNVDAIL EEYANCKKSQ GNVDNKEYAV NEVVAGIKEY FNVMLGTQLL YKFERPQYAE ILLAHPDAPM SQVYGAPHLL RLFVRIGAML AYTPLDEKSL ALLLGYLHDF LKYLAKNSAS LFTASDYKVA SAEYHRKAL (1-288 aa encoded by BC056899 )
MORF4L2 is a protein coding gene. Component of the NuA4 histone acetyltransferase complex which is involved in transcriptional activation of select genes principally by acetylation of nucleosomal histone H4 and H2A. This modification may both alter nucleosome - DNA interactions and promote interaction of the modified histones with other proteins which positively regulate transcription. This complex may be required for the activation of transcriptional programs associated with oncogene and proto-oncogene mediated growth induction, tumor suppressor mediated growth arrest and replicative senescence, apoptosis, and DNA repair. The NuA4 complex ATPase and helicase activities seem to be, at least in part, contributed by the association of RUVBL1 and RUVBL2 with EP400. NuA4 may also play a direct role in DNA repair when directly recruited to sites of DNA damage. Also component of the MSIN3A complex which acts to repress transcription by deacetylation of nucleosomal histones.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Liver regeneration-related protein LRRG00119; MORF-related gene X; MORF-related gene X protein; Mortality factor 4-like protein 2; MSL3-2 protein; OTTHUMP00000023756; Protein MSL3-2; RP5-1055C14.2; Sid 393; Transcription factor-like protein MRGX
Gene Aliases: 2410017O14Rik; KIAA0026; LRRG00119; mKIAA0026; MORF4L2; MORFL2; MRGX; Sid393; Sid393p
UniProt ID: (Human) Q15014, (Mouse) Q9R0Q4, (Rat) Q6QI89
Entrez Gene ID: (Human) 9643, (Mouse) 56397, (Rat) 317413
Molecular Function:
chromatin/chromatin-binding, or -regulatory protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support