Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: DINDKSITDS TVRVIWAYHH EDAGEAGPKY HDSNRGTKSL RLLNPEKTSV LSTALPYFDL VNQDVPIPNK DTTYWCQMFK IPVFQEKHHV IKVEPV
Highest antigen sequence identity to the following orthologs: Mouse - 84%, Rat - 90%.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: DBH-like monooxygenase protein 1; dopamine-oxygenase; Monooxygenase X; monooxygenase, DBH-like 1
Gene Aliases: dJ248E1.1; MOX; MOXD1; PRO5780; UNQ2493/PRO5780
UniProt ID: (Human) Q6UVY6
Entrez Gene ID: (Human) 26002
Molecular Function:
hydroxylase
oxidoreductase
metabolite interconversion enzyme
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support