Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 24
Antibody detects endogenous levels of total MPV17.
This gene encodes a mitochondrial inner membrane protein that is implicated in the metabolism of reactive oxygen species. Mutations in this gene have been associated with the hepatocerebral form of mitochondrial DNA depletion syndrome.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: h-MPV; hMPV; Mpv-17; MpV17 mitochondrial inner membrane protein; Mpv17 protein; Mpv17 transgene, kidney disease mutant-like; Mpv17, human homolog of glomerulosclerosis and nephrotic syndrome; OTTHUMP00000122591; Protein Mpv17
Gene Aliases: MPV17; Mpv17l; MTDPS6; SYM1; Tg.Mpv17
UniProt ID: (Human) P39210, (Mouse) P19258, (Rat) Q5BK62
Entrez Gene ID: (Human) 4358, (Mouse) 17527, (Rat) 360463
Molecular Function:
transporter
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support