Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
The antibody can recognize the isoform 1, isoform 2, and isoform 4.
MRAP encodes a melanocortin receptor-interacting protein. The encoded protein regulates trafficking and function of the melanocortin 2 receptor in the adrenal gland. The encoded protein can also modulate signaling of other melanocortin receptors. Mutations in this gene have been associated with familial glucocorticoid deficiency type 2.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: B27; Fat cell-specific low molecular weight protein; Fat tissue-specific low MW protein; Melanocortin-2 receptor accessory protein
Gene Aliases: 1110025G12Rik; B27; C21orf61; FALP; FGD2; GCCD2; MRAP; ORF61
UniProt ID: (Human) Q8TCY5, (Mouse) Q9D159
Entrez Gene ID: (Human) 56246, (Mouse) 77037
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support