Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
Additional Information: This antibody is expected to recognise isoform 1 (NP_006782.1) and isoform 2 (NP_996670.1).
This antibody is tested in Peptide ELISA: antibody detection limit dilution 1:128,000.
MRG15 ia a component of the NuA4 histone acetyltransferase (HAT) complex which is involved in transcriptional activation of select genes principally by acetylation of nucleosomal histone H4 and H2A. This modification may both alter nucleosome-DNA interactions and promote interaction of the modified histones with other proteins which positively regulate transcription. This complex may be required for the activation of transcriptional programs associated with oncogene and proto-oncogene mediated growth induction, tumor suppressor mediated growth arrest and replicative senescence, apoptosis and DNA repair.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: MGC10631; MORF-related gene 15 protein; Mortality factor 4-like protein 1; testis expressed gene 189; Testis-expressed gene 189 protein; Transcription factor-like protein MRG15
Gene Aliases: mKIAA4002; Morf4l1; MORFRG15; MRG15; TEG-189; Tex189
UniProt ID: (Rat) Q6AYU1, (Mouse) P60762
Entrez Gene ID: (Rat) 300891, (Mouse) 21761
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support