Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Antibody detects endogenous levels of total MRGRG.
Orphan receptor. May regulate nociceptor function and/or development, including the sensation or modulation of pain.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: G protein-coupled receptor 169; G protein-coupled receptor MRGG; G-protein coupled receptor 169; Mas-related G-protein coupled receptor member G; MAS-related GPR, member G
Gene Aliases: GPR169; MRGG; MRGPRG
UniProt ID: (Human) Q86SM5
Entrez Gene ID: (Human) 386746
Molecular Function:
G-protein coupled receptor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support