Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
For reconstitution, we recommend adding 100 µL distilled water to a final antibody concentration of about 1 mg/mL. To use this carrier-free antibody for conjugation experiments, we strongly recommend performing another round of desalting. (Zeba Spin Desalting Columns, 7KMWCO, 0.5 mL, Product # 89882)
MGC3207 catalyzes the interconversion of methylthioribose-1-phosphate (MTR-1-P) into methylthioribulose-1-phosphate (MTRu-1-P). Independently from catalytic activity, MGC3207 promotes cell invasion in response to constitutive RhoA activation by promoting FAK tyrosine phosphorylation and stress fiber turnover.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: M1Pi; Mediator of RhoA-dependent invasion; Methylthioribose-1-phosphate isomerase; methylthioribose-1-phosphate isomerase homolog; MGC3207; MRI1; MTR-1-P isomerase; S-methyl-5-thioribose-1-phosphate isomerase; S-methyl-5-thioribose-1-phosphate isomerase 1; Translation initiation factor eIF-2B subunit alpha/beta/delta-like protein
Gene Aliases: MRDI; MRI1; MTNA; UNQ6390/PRO21135; Ypr118w
UniProt ID: (Human) Q9BV20
Entrez Gene ID: (Human) 84245
Molecular Function:
isomerase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support