Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MLPAAARPLW GPCLGLRAAA FRLARRQVPC VCAVRHMRSS GHQRCEALAG APLDNAPKEY PPKIQQLVQD IASLTLLEIS DLNELLKKTL KIQDVGLVPM GGVMSGAVPA AAAQEAVEED IPIAKERTHF TVRLTEAKPV DKVKLIKEIK NYIQGINLVQ AKKLVESLPQ EIKANVAKAE AEKIKAALEA VGGTVVLE
MRPL12 (mitochondrial ribosomal protein L12), also referred to as 5c5-2, L12mt, MRPL7 or RPML12, is a mammalian mitochondrial ribosomal protein that is involved in protein synthesis within the mitochondrion. MRPL12 is enhanced in growth-stimulated cells as a result of transcriptional activation, suggesting that it may function as a translational regulator of mitochondrial mRNAs. Impairment of MRPL12 leads to reduction in cell growth rate, decreased mitochondrial ATP production and abolition of mitochondrial oxidative phosphorylation. MRPL12 is cleaved during its translocation across the mitochondrial membrane and it exists as dimers that bind the large ribosomal subunit. MRPL12 is 198 amino acids in length, belongs to the ribosomal protein L12P family and is highly expressed in the colon.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 39S ribosomal protein L12, mitochondrial; 5c5-2; FLJ60124; L12mt; Large ribosomal subunit protein bL12m; MGC8610; MRP-L12
Gene Aliases: 5c5-2; L12mt; MRP-L31/34; MRPL12; MRPL7; MRPL7/L12; RPML12
UniProt ID: (Human) P52815
Entrez Gene ID: (Human) 6182
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support