Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MSSFSRAPQ QWATFARIWY LLDGKMQPPG KLAAMASIRL QGLHKPVYHA LSDCGDHVVI MNTRHIAFSG NKWEQKVYSS HTGYPGGFRQ VTAAQLHLRD PVAIVKLAIY GMLPKNLHRR TMMERLHLFP DEYIPEDILK NLVEELPQPR KIPKRLDEYT QEEIDAFPRL WTPPEDYRL (1-178 aa encoded by BC009190 )
Mammalian mitochondrial ribosomal proteins are encoded by nuclear genes and help in protein synthesis within the mitochondrion. Mitochondrial ribosomes (mitoribosomes) consist of a small 28S subunit and a large 39S subunit. They have an estimated 75% protein to rRNA composition compared to prokaryotic ribosomes, where this ratio is reversed. Another difference between mammalian mitoribosomes and prokaryotic ribosomes is that the latter contain a 5S rRNA. Among different species, the proteins comprising the mitoribosome differ greatly in sequence, and sometimes in biochemical properties, which prevents easy recognition by sequence homology. This gene encodes a 39S subunit protein.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 39S ribosomal protein L13, mitochondrial; L13mt; Large ribosomal subunit protein uL13m; MRP-L13
Gene Aliases: 1110002D09Rik; L13; L13A; L13mt; MRPL13; RPL13; RPML13
UniProt ID: (Human) Q9BYD1, (Mouse) Q9D1P0
Entrez Gene ID: (Human) 28998, (Mouse) 68537, (Rat) 299938
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support