Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: IDKEYYRKP VEELTEEEKY VRELKKTQLI KAAPAGKTSS VFEDPVISKF TNMMMIGGNK VLARSLMIQT LEAVKRKQFE KYHAASAEEQ ATIERNPYTI FHQALKNCEP MIGLVPILKG GRFYQVPVPL PDRRRRFLAM KWMITECRDK KHQRTLMPEK LSHKLLEAFH NQGPVIKRKH DLHKMAEANR ALAHYRWW (46-242 aa encoded by BC000241 )
Mammalian mitochondrial ribosomal proteins are encoded by nuclear genes and help in protein synthesis within the mitochondrion. Mitochondrial ribosomes (mitoribosomes) consist of a small 28S subunit and a large 39S subunit. They have an estimated 75% protein to rRNA composition compared to prokaryotic ribosomes, where this ratio is reversed. Another difference between mammalian mitoribosomes and prokaryotic ribosomes is that the latter contain a 5S rRNA. Among different species, the proteins comprising the mitoribosome differ greatly in sequence, and sometimes in biochemical properties, which prevents easy recognition by sequence homology. This gene encodes a 28S subunit protein. In the prokaryotic ribosome, the comparable protein is thought to play an essential role in organizing the 3' domain of the 16 S rRNA in the vicinity of the P- and A-sites. Pseudogenes corresponding to this gene are found on chromosomes 8p and 12p.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 28S ribosomal protein S7, mitochondrial; 30S ribosomal protein S7 homolog; bMRP-27a; bMRP27a; MRP-S7; Small ribosomal subunit protein uS7m
Gene Aliases: bMRP27a; MRP-S; MRP-S7; MRPS7; RP-S7; RPMS7; S7mt
UniProt ID: (Human) Q9Y2R9
Entrez Gene ID: (Human) 51081
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support