Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
Immunogen sequence: MALGLKCFRM VHPTFRNYLA ASIRPVSEVT LKTVHERQHG HRQYMAYSAV PVRHFATKKA KAKGKGQSQT RVNINAALVE DIINLEEVNE EMKSVIEALK DNFNKTLNIR TSPGSLDKIA VVTADGKLAL NQISQISMKS PQLILVNMAS FPECTAAAIK AIRESGMNLN PEVEGTLIRV PIPQVTREHR EMLVKLAKQN TNKAKDSLRK VRTNSMNKLK KSKDTVSEDT IRLIEKQISQ MADDTVAELD RHLAVKTKEL LG
Responsible for the release of ribosomes from messenger RNA at the termination of protein biosynthesis. May increase the efficiency of translation by recycling ribosomes from one round of translation to another. Ota T., Nat. Genet. 36:40-45(2004). Humphray S.J., Nature 429:369-374(2004). Zhang Y., Biochim. Biophys. Acta 1443:245-250(1998).
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Ribosome-recycling factor, mitochondrial; Ribosome-releasing factor, mitochondrial; RRF
Gene Aliases: MRFF; MRRF; MTRRF; RRF
UniProt ID: (Human) Q96E11
Entrez Gene ID: (Human) 92399
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support