Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
Immunogen sequence: MSTGSVSDPE EMELRGLQRE YPVPASKRPP LRGVERSYAS PSDNSSAEEE DPDGEEERCA LGTAGSAEGC KRKRPRVAGG GGAGGSAGGG GKKPLPAKGS
MyoD-E or musculin-E heterodimers bind avidly to consensus E box motifs, which are functionally important elements in the promoter regions of many musclespecific terminal differentiation genes. MyoD complexes potently induce transcriptional activation, while musculin complexes bind adjacent to MyoD DNA-binding regions to represses MyoD activity, which then results in the delayed expression of muscle-specific genes. Musculin is highly expressed in undifferentiated and proliferating myoblasts in culture, and its expression is down regulated during myogenesis and at the onset of terminal differentiation.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ABF-1; Activated B-cell factor 1; activated B-cell factor 1, homolog of mouse musculin; activated B-cell factor-1; bHLHa22; Class A basic helix-loop-helix protein 22; Musculin
Gene Aliases: ABF-1; ABF1; BHLHA22; MSC; MYOR
UniProt ID: (Human) O60682
Entrez Gene ID: (Human) 9242
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support