Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
This Antibody was verified by Knockout to ensure that the antibody binds to the antigen stated.
FIGURE: 1 / 18
Positive Control: Human skin tissue lysate, rat brain tissue lysate, human thyroid tissue, human colon carcinoma tissue, human skin tissue, human breast carcinoma tissue, human esophagus tissue, human placenta tissue, mouse testis tissue, mouse colon tissue, mouse placenta tissue, human gastric adenocarcinoma tissue, mouse spleen tissue, mouse small intestine tissue, K562.
Subcellular Location: Nucleus, Chromosome.
The DNA mismatch repair MSH2/MSH6 heterodimer is responsible for the detection of DNA damage, and may promote apoptosis in normal cells, thus protecting mammals from ultraviolet-induced malignant transformation and hence the prevention of skin cancer.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: DNA mismatch repair protein Msh6; G/T mismatch-binding protein; GTBP; hMSH6; mutS (E. coli) homolog 6; MutS alpha 160kDa subunit; mutS homolog 6; MutS protein homolog 6; MutS-alpha 160 kDa subunit; MutS-alpha 160kDa subunit; p160; rCG61559-like; Sperm associated protein; sperm-associated protein
Gene Aliases: AU044881; AW550279; GTBP; GTMBP; HNPCC5; HSAP; MSH6; p160
UniProt ID: (Human) P52701, (Mouse) P54276
Entrez Gene ID: (Rat) 100360342, (Human) 2956, (Mouse) 17688
Molecular Function:
DNA metabolism protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support