Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 5
Positive control: A549 cell lysate, HepG2 cell lysate, human tonsil tissue, human colon carcinoma tissue, human stomach carcinoma tissue, mouse small intestine tissue.
Predicted band size: 153 kDa
Subcellular Location: Nucleus, Chromosome.
The DNA mismatch repair MSH2/MSH6 heterodimer is responsible for the detection of DNA damage, and may promote apoptosis in normal cells, thus protecting mammals from ultraviolet-induced malignant transformation and hence the prevention of skin cancer.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: DNA mismatch repair protein Msh6; G/T mismatch-binding protein; GTBP; hMSH6; mutS (E. coli) homolog 6; MutS alpha 160kDa subunit; MutS protein homolog 6; MutS-alpha 160 kDa subunit; MutS-alpha 160kDa subunit; p160; Sperm associated protein; sperm-associated protein
Gene Aliases: AU044881; AW550279; GTBP; GTMBP; HNPCC5; HSAP; MSH6; p160
UniProt ID: (Human) P52701, (Mouse) P54276
Entrez Gene ID: (Human) 2956, (Mouse) 17688
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support