Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: AKQMEAALSSK ENYQKVLSEH GFGPITTDIR EGQTFYYAED YHQQYLSKNP NGYCGLGGTG VSCPVGIK
Highest antigen sequence identity to the following orthologs - mouse 87%, rat 65%.
Methionine sulfoxide reductase (MsrA) reduces methionine sulfoxide (MetO) residues in proteins and free MetO to Methionine (Met). The catalytic activity of MsrA is dependent of bound metal and cofactors but it requires reducing equivalents from either DTT or a thioredoxin-regenerating system. MsrA plays an essential role in protecting cells against oxidative damage. The substrates of MsrA include calmodulin, HIV protease and a 1-proteinase-inhibitor. Recent studies indicate that there is a connection between MsrA and Alzheimer's disease in mammals.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: cytosolic methionine-S-sulfoxide reductase; Mitochondrial peptide methionine sulfoxide reductase; peptide met (O) reductase; Peptide Met(O) reductase; Peptide-methionine (S)-S-oxide reductase; PMSR; Protein-methionine-S-oxide reductase
Gene Aliases: MSRA; PMSR
UniProt ID: (Human) Q9UJ68
Entrez Gene ID: (Human) 4482
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support