Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
MSRB2 catalyzes the reduction of free and protein-bound methionine sulfoxide to methionine.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Methionine-R-sulfoxide reductase B2, mitochondrial; MsrB2; pilin-like transcription factor
Gene Aliases: 2310050L06Rik; CBS-1; CBS1; CGI-131; Mrsb; MSRB; MSRB2; PILB
UniProt ID: (Human) Q9Y3D2, (Rat) Q4FZX5, (Mouse) Q78J03
Entrez Gene ID: (Human) 22921, (Rat) 361286, (Mouse) 76467
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support