Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MAPAADMTS LPLGVKVEDS AFGKPAGGGA GQAPSAAAAT AAAMGADEEG AKPKVSPSLL PFSVEALMAD HRKPGAKESA LAPSEGVQAA GGSAQPLGVP PGSLGAPDAP SSPRPLGHFS VGGLLKLPED ALVKAESPEK PERTPWMQSP RFSPPPARRL SPPACTLRKH KTNRKPRTPF TTAQLLALER KFRQKQYLSI AERAEFSSSL SLTETQVKIW FQNRRAKAKR LQEAELEKLK MAAKPMLPPA AFGLSFPLGG PAAVAAAAGA SLYGASGPFQ RAALPVAPVG LYTAHVGYSM YHLT (1-303 aa encoded by BC021285)
MSX1 is a member of the muscle segment homeobox gene family. It functions as a transcriptional repressor during embryogenesis through interactions with components of the core transcription complex and other homeoproteins. MSX1 may also have roles in limb-pattern formation, craniofacial development, particularly odontogenesis, and tumor growth inhibition. Mutations in the gene, which was once known as homeobox 7, have been associated with nonsyndromic cleft lip with or without cleft palate 5, Witkop syndrome, Wolf-Hirschom syndrome, and autosomoal dominant hypodontia.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: homeo box, msh-like 1; homeobox 7; Homeobox protein Hox-7; Homeobox protein MSX-1; homeobox, msh-like 1; Hox-7.1; msh homeo box 1; Msh homeobox 1-like protein; msh homeobox homolog 1; muscle-segment homeobox
Gene Aliases: AA675338; AI324650; ECTD3; Hox-7; HOX7; Hox7.1; HYD1; msh; MSX1; STHAG1
UniProt ID: (Human) P28360, (Mouse) P13297
Entrez Gene ID: (Human) 4487, (Mouse) 17701, (Rat) 81710
Molecular Function:
homeodomain transcription factor
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support