Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
Immunogen sequence: MASPSKGNDL FSPDEEGPAV VAGPGPGPGG AEGAAEERRV KVSSLPFSVE ALMSDKKPPK EASPLPAESA SAGATLRPLL LSGHGAREAH SPGPLVKPFE TASVKSENSE DGAAWMQEPG RYSPPPRHTS PTT
Msx2 is a transcriptional repressor whose normal activity may establish a balance between survival and apoptosis of neural crest-derived cells required for proper craniofacial morphogenesis. This protein may also have a role in promoting cell growth under certain conditions and may be an important target for the RAS signaling pathways. Mutations in the Msx2 gene are associated with parietal foramina 1 and craniosynostosis type 2.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Homeobox protein Hox-8; Homeobox protein MSX-2; msh homeo box 2; msh homeobox homolog 2
Gene Aliases: CRS2; FPP; HOX8; MSH; MSX2; PFM; PFM1
UniProt ID: (Human) P35548
Entrez Gene ID: (Human) 4488
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support