Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
NeoBiotechnologies
Immunohistochemistry (PFA fixed): incubate antibody for 30 min RT. Staining of formalin-fixed tissues requires heating tissue sections in 10mM Tris with 1mM EDTA, pH 9.0, for 45 min at 95 degrees C followed by cooling at RT for 20 minutes.
This gene encodes an enzyme that plays a major role in polyamine metabolism and is important for the salvage of both adenine and methionine. The encoded enzyme is deficient in many cancers because this gene and the tumor suppressor p16 gene are co-deleted. Multiple alternatively spliced transcript variants have been described for this gene, but their full-length natures remain unknown.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 5'-methylthioadenosine phosphorylase; epididymis luminal protein 249; MeSAdo phosphorylase; MTA phosphorylase; MTAP; MTAPase; S-methyl-5'-thioadenosine phosphorylase
Gene Aliases: BDMF; c86fus; DMSFH; DMSMFH; HEL-249; LGMBF; MSAP; MTAP
UniProt ID: (Human) Q13126
Entrez Gene ID: (Human) 4507
Molecular Function:
nucleotide kinase
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support