Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Sequence of this protein is as follows: MAPAEILNGK EISAQIRARL KNQVTQLKEQ VPGFTPRLAI LQVGNRDDSN LYINVKLKAA EEIGIKATHI KLPRTTTESE VMKYITSLNE DSTVHGFLVQ LPLDSENSIN TEEVINAIAP EKDVDGLTSI NAGRLARGDL NDCFIPCTPK GCLELIKETG VPIAGRHAVV VGRSKIVGAP MHDLLLWNNA TVTTCHSKTA HLDEEVNKGD ILVVATGQPE MVKGEWIKPG AIVIDCGINY VPDDKKPNGR KVVGDVAYDE AKERASFITP VPGGVGPMTV AMLMQSTVES AKRFLEKFKP GKWMIQYNNL NLKTPVPSDI DISRSCKPKP IGKLAREIGL LSEEVELYGE TKAKVLLSAL ERLKHRPDGK YVVVTGITPT PLGEGKSTTT IGLVQALGAH LYQNVFACVR QPSQGPTFGI KGGAAGGGYS QVIPMEEFNL HLTGDIHAIT AANNLVAAAI DARIFHELTQ TDKALFNRLV PSVNGVRRFS DIQIRRLKRL GIEKTDPTTL TDEEINRFAR LDIDPETITW QRVLDTNDRF LRKITIGQAP TEKGHTRTAQ FDISVASEIM AVLALTTSLE DMRERLGKMV VASSKKGEPV SAEDLGVSGA LTVLMKDAIK PNLMQTLEGT PVFVHAGPFA NIAHGNSSII ADRIALKLVG PEGFVVTEAG FGADIGMEKF FNIKCRYSGL CPHVVVLVAT VRALKMHGGG PTVTAGLPLP KAYIQENLEL VEKGFSNLKK QIENARMFGI PVVVAVNAFK TDTESELDFI SRLSREHGAF DAVKCTHWAE GGKGALALAQ AVQRAAQAPS SFQLLYDLKL PVEDKIRIIA QKIYGADDIE LLPEAQHKAE VYTKQGFGNL PICMAKTHLS LSHNPEQKGV PTGFILPIRD IRASVGAGFL YPLVGTMSTM PGLPTRPCFY DIDLDPETEQ VNGLF
This gene encodes a protein that possesses three distinct enzymatic activities, 5,10-methylenetetrahydrofolate dehydrogenase, 5,10-methenyltetrahydrofolate cyclohydrolase and 10-formyltetrahydrofolate synthetase. Each of these activities catalyzes one of three sequential reactions in the interconversion of 1-carbon derivatives of tetrahydrofolate, which are substrates for methionine, thymidylate, and de novo purine syntheses. The trifunctional enzymatic activities are conferred by two major domains, an aminoterminal portion containing the dehydrogenase and cyclohydrolase activities and a larger synthetase domain.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 5,10-methylenetetrahydrofolate dehydrogenase, 5,10-methylenetetrahydrofolate cyclohydrolase, 10-formyltetrahydrofolate synthetase; C-1-tetrahydrofolate synthase, cytoplasmic; C1-THF synthase; cytoplasmic C-1-tetrahydrofolate synthase; Epididymis secretory sperm binding protein; methenyltetrahydrofolate; methylenetetrahydrofolate dehydrogenase (NADP+ dependent) 1, methenyltetrahydrofolate cyclohydrolase, formyltetrahydrofolate synthetase
Gene Aliases: MTHFC; MTHFD; MTHFD1
UniProt ID: (Human) P11586
Entrez Gene ID: (Human) 4522
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support