Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: TNDRFLRKIT IGQAPTEKGH TRTAQFDISV ASEIMAVLAL TTSLEDMRER LGKMVVASSK KGEPVSAEDL GVSGALTVLM KDAIKPNLMQ TLEGTPVFVH AGPFANIAHG NSSIIADQIA LKLVGPEGFV VTEAGFGADI GMEKFFNIKC RYSGLCPHVV VLVATVRALK MHGGGPTVTA GLPLPKAYIQ ENLELVEKGF SNLKKQIENA RMFGIPVVVA VNAFKTDTES ELDLISRLSR EHGAFDAVKC THWAEGGKGA LALAQAVQRA AQAPSSFQLL YDLKLPVEDK IRIIAQKIYG ADDIELLPEA QHKAEVYTKQ GFGNLPICMA KTHLSLSHNP EQKGVPTGFI LPIRDIRASV GAGFLYPLVG TMSTMPGLPT RPCFYDIDLD PETEQVNGLF
This gene encodes a protein that possesses three distinct enzymatic activities, 5,10-methylenetetrahydrofolate dehydrogenase, 5,10-methenyltetrahydrofolate cyclohydrolase and 10-formyltetrahydrofolate synthetase. Each of these activities catalyzes one of three sequential reactions in the interconversion of 1-carbon derivatives of tetrahydrofolate, which are substrates for methionine, thymidylate, and de novo purine syntheses. The trifunctional enzymatic activities are conferred by two major domains, an aminoterminal portion containing the dehydrogenase and cyclohydrolase activities and a larger synthetase domain.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 5,10-methylenetetrahydrofolate dehydrogenase, 5,10-methylenetetrahydrofolate cyclohydrolase, 10-formyltetrahydrofolate synthetase; C-1-tetrahydrofolate synthase, cytoplasmic; C1-tetrahydrofolate synthase; C1-THF synthase; cytoplasmic C-1-tetrahydrofolate synthase; Epididymis secretory sperm binding protein; methenyltetrahydrofolate; methylenetetrahydrofolate dehydrogenase (NADP+ dependent) 1, methenyltetrahydrofolate cyclohydrolase, formyltetrahydrofolate synthetase; methylenetetrahydrofolate dehydrogenase (NADP+ dependent), methenyltetrahydrofolate cyclohydrolase, formyltetrahydrofolate synthase
Gene Aliases: Dcs; E430024A07Rik; MTHFC; MTHFD; MTHFD1
UniProt ID: (Human) P11586, (Mouse) Q922D8, (Rat) P27653
Entrez Gene ID: (Human) 4522, (Mouse) 108156, (Rat) 64300
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support