Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Cell lysate).
Immunogen sequence: VFKTDTRAEI DLVCELAKRA GAFDAVPCYH WSVGGKGSVD LARAVREAAS KRSRFQFLYD VQVPIVDKIR TIAQAVYGAK DIELSPEAQA KIDRYTQQG
One-carbon substituted forms of tetrahydrofolate (THF) are involved in the de novo synthesis of purines and thymidylate and support cellular methylation reactions through the regeneration of methionine from homocysteine. MTHFD1L is an enzyme involved in THF synthesis in mitochondria (Christensen et al., 2005 [PubMed 15611115]).
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 10-formyl-THF synthetase; Formyltetrahydrofolate synthetase; formyltetrahydrofolate synthetase domain containing 1; Monofunctional C1-tetrahydrofolate synthase, mitochondrial
Gene Aliases: dJ292B18.2; FTHFSDC1; MTC1THFS; MTHFD1L
UniProt ID: (Human) Q6UB35
Entrez Gene ID: (Human) 25902, (Rat) 361472
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support