Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MAALFLKRL TLQTVKSENS CIRCFGKHIL QKTAPAQLSP IASAPRLSFL IHAKAFSTAE DTQNEGKKIK KNKTAFSNVG RKISQRVIHL FDEKGNDLGN MHRANVIRLM DERDLRLVQR NTSTEPAEYQ LMTGLQILQE RQRLREMEKA NPKTGPTLRK ELILSSNIGQ HDLDTKTKQI QQWIKKKHLV QITIKKGKNV DVSENEMEEI FHQILQTMPG IATFSSRPQA VQGGKALMCV LRALSKNEEK AYKETQETQE RDTLNKDHGN DKESNVLHQ (1-278 aa encoded by BC046166 )
This gene encodes a translation initiation factor that is involved in mitochondrial protein synthesis. Polymorphism in this gene is associated with the onset of Parkinson's disease. Alternate splicing results in multiple transcript variants. A pseudogene of this gene is found on chromosome 5.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: IF-3(Mt); IF-3Mt; IF3(mt); IF3mt; Translation initiation factor IF-3, mitochondrial
Gene Aliases: 2810012L14Rik; AI414549; DC38; IF3mt; MTIF3
UniProt ID: (Human) Q9H2K0, (Mouse) Q9CZD5
Entrez Gene ID: (Human) 219402, (Mouse) 76366
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support