Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Zeta
This product is diluted and in a ready-to-use formulation.
A recommended positive control tissue for this product is Colon carcinoma, however positive controls are not limited to this tissue type.
Secreted epithelial mucins are large macromolecules which exhibit extreme polydispersity. Mucin 2 is the major intestinal mucin. O-glycans are attached to MUC2 in a potentially diverse arrangement, which is crucial for their interaction with endogeneous and exogeneous lectins.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Intestinal mucin-2; MUC-2; mucin 2, intestinal/tracheal; Mucin-2; mucin-like protein
Gene Aliases: MLP; MUC-2; MUC2; SMUC
UniProt ID: (Human) Q02817
Entrez Gene ID: (Human) 4583
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support