Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 3
Positive test controls include: HeLa.
Immunogen sequence: WATVSVIALS NILHASASLP PEYQNRTEGL LGVWNNNPED DFRMPNGSTI PPGSPEEMLF HFGMTWQING TGLLGKRNDQ LPSNFTPVFY SQLQKNSSWA EHLISNCDGD
Nearly all mucosal surfaces are protected by mucin transmembrane glycoproteins, from bronchioles to corneas to reproductive tracts. Mucin 4 (MUC4) is membrane-bound and is important in protecting and lubricating. MUC4 expression is erroneous in many carcinomas and sarcomas, including fibromyoxoid sarcoma and pancreatic adenocarcinoma. Immunohistochemical staining for MUC4 has been discussed as a potential biomarker for detection of these cancers. MUC4 seems to be most predominantly affected in Asian diseases - Taiwanese infertility and endometriosis, as well as gallstone formation in Chinese men, are connected to MUC4, where similar diseases in other populations do not share these MUC4 mutations. Because MUC4 is so important in mucosal surface protection, it is not surprising that it has been implicated as failing in many metastatic cancer invasions.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Ascites sialoglycoprotein; ASGP; ASGP1; MUC-4; mucin 4, tracheobronchial; Mucin-4; Mucin4; Pancreatic adenocarcinoma mucin; pre-sialomucin complex; sialomucin ascites sialoglycoprotein-1; sialomucin complex; Testis mucin; Tracheobronchial mucin
Gene Aliases: ASGP; ASGP-1; HSA276359; MUC-4; MUC4; Psmc
UniProt ID: (Human) Q99102
Entrez Gene ID: (Human) 4585, (Rat) 303887
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support