Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MLPDRSRAA RDRANQKLVE YIVKAKGAES HLRAILKSRK PSRWLQTFLS SSQYVTCVET YLEDEGQLDL VVKYLQGVYQ EVGAKVLQRT NGDRIRFILD VLLPEAIICA ISAVDEVDYK TAEEKYIKGP SLSYREKEIF DNQLLEERNR RRR (1-152 aa encoded by BC008098)
Mutated melanoma-associated antigen 1 (MUM1) is a 78 kDa protein encoded by the MUM1 gene. MUM1 is expressed in a subset of B cells in the light zone of the germinal center, plasma cells, activated T cells and a wide spectrum of related hematolymphoid neoplasms. Antibodies to MUM1 are useful in the sub classification of lymphoid malignancies.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: FLJ14868; FLJ22283; melanoma ubiquitous mutated protein; MGC131891; MGC163315; MUM-1; Mutated melanoma-associated antigen 1; Protein expandere; PWWP domain-containing DNA repair factor 3A; PWWP domain-containing protein MUM1; PWWP3A
Gene Aliases: EXPAND1; HSPC211; MUM-1; MUM1; PWWP3A
UniProt ID: (Human) Q2TAK8
Entrez Gene ID: (Human) 84939
Molecular Function:
DNA metabolism protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support