Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
The recommended shelf life for this product is 1 year from date of receipt.
MUPP1 (multi PDZ domain protein 1, MPDZ) was discovered as an interacting partner with the 5-HT2C receptor1, and was found to co-localize with 5-HT2A or 5-HT2C receptors in all regions of the mouse brain, including the choroid plexus. MUPP1 contains 13 PDZ domains but no apparent catalytic domain. MUPP1 is concentrated in tight junctions of polarized epithelial cells and binds to claudin-1 and junctional adhesion molecule (JAM). MUPP1 may therefore function as a multivalent scaffold protein that recruits various proteins to tight junctions. MUPP1 also interacts with claudin-8 and may play a role in tight junction barrier function.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: DKFZp781P216; FLJ25909; FLJ34626; FLJ90240; Multi-PDZ domain protein 1; Multiple PDZ domain protein
Gene Aliases: HYC2; MPDZ; MUPP1
UniProt ID: (Human) O75970
Entrez Gene ID: (Human) 8777
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support