Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
Sequence of this protein is as follows: MTPLVSRLSR LWAIMRKPRA AVGSGHRKQA ASQEGRQKHA KNNSQAKPSA CDGLARQPEE VVLQASVSSY HLFRDVAEVT AFRGSLLSWY DQEKRDLPWR RRAEDEMDLD RRAYAVWVSE VMLQQTQVAT VINYYTGWMQ KWPTLQDLAS ASLEEVNQLW AGLGYYSRGR RLQEGARKVV EELGGHMPRT AETLQQLLPG VGRYTAGAIA SIAFGQATGV VDGNVARVLC RVRAIGADPS STLVSQQLWG LAQQLVDPAR PGDFNQAAME LGATVCTPQR PLCSQCPVES LCRARQRVEQ EQLLASGSLS GSPDVEECAP NTGQCHLCLP PSEPWDQTLG VVNFPRKASR KPPREESSAT CVLEQPGALG AQILLVQRPN SGLLAGLWEF PSVTWEPSEQ LQRKALLQEL QRWAGPLPAT HLRHLGEVVH TFSHIKLTYQ VYGLALEGQT PVTTVPPGAR WLTQEEFHTA AVSTAMKKVF RVYQGQQPGT CMGSKRSQVS SPCSRKKPRM GQQVLDNFFR SHISTDAHSL NSAAQ
MYH (mutY homolog (E. coli )) is a DNA glycosylase mismatch repair enzyme that in conjunction with mutM (OGG1), cleaves adenine residues paired with either oxidized (8-hydroxyguanines) or non-modified guanines in order to correct A/G and A/C mismatches. Repair of most modified and mispaired bases in the genome is initiated by DNA glycosylases, which bind and cleave N-glycosyl bonds to initiate base excision repair. MYH is crucial for the avoidance of mutations resulting from oxidative DNA damage. Multiple N-terminal splice variants of MYH exist in mammalian cells. Increasing levels of MYH in A549 cells exposed to oxygen and infrared radiation leads to improvements in cell survival. Biallelic MYH germ-line mutations predispose humans to colorectal adenomas and carcinomas. MYH is abundant in neurons where mitochondrial genomes exposed to reactive oxygen species (ROS) that damage DNA must maintain integrity over the entire mammalian life span.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: A/G-specific adenine DNA glycosylase; Adenine DNA glycosylase; hMYH; MGC4416; MutY homolog; mutY-like protein; OTTHUMP00000009098; OTTHUMP00000009102; RP4-534D1.2
Gene Aliases: MUTYH; MYH
UniProt ID: (Human) Q9UIF7
Entrez Gene ID: (Human) 4595
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support