Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MLSEVLLVSA PGKVILHGEH AVVHGKVALA VSLNLRTFLR LQPHSNGKVD LSLPNIGIKR AWDVARLQSL DTSFLEQGDV TTPTSEQVEK LKEVAGLPDD CAVTERLAVL AFLYLYLSIC RKQRALPSLD IVVWSELPPG AGLGSSAAYS VCLAAALLTV CEEIPNPLKD GDCVNRWTKE DLELINKWAF QGERMIHGNP SGVDNAVSTW GGALRYHQGK ISSLKRSPAL QILLTNTKVP RNTRALVAGV RNRLLKFPEI VAPLLTSIDA ISLECERVLG EMGEAPAPEQ YLVLEELIDM NQHHLNALGV GHASLDQLCQ VTRARGLHSK LTGAGGGGCG ITLLKPGLEQ PEVEATKQAL TSCGFDCLET SIGAPGVSIH SATSLDSRVQ QALDGL
Mevalonate kinase (MVK) is an early enzyme in isoprenoid and sterol synthesis. Mevalonate kinase catalyzes the ATP-dependent phosphorylation of mevalonic acid to form mevalonate 5-phosphate. Deficiency in MVK activity contributes to mevalonic aciduria and hyperimmunoglobulinemia D/periodic fever syndrome (HIDS).
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: KIME; LH receptor mRNA-binding protein; Mevalonate kinase; mevalonate kinase 1; MK
Gene Aliases: LRBP; MK; MVK; MVLK; POROK3
UniProt ID: (Human) Q03426
Entrez Gene ID: (Human) 4598
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support