Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Cell lysate).
Immunogen sequence: MATEEFIIRI PPYHYIHVLD QNSNVSRVEV GPKTYIRQDN ERVLFAPMRM VTVPPRHYCT VANPVSRDAQ GLVLFDVTGQ VRLRHADLEI RLAQDPFPLY PGEVLEKDIT PLQVVLPNTA LHLKALLDFE DKDGDKVVAG DEWLFEGPGT YIPRKEVEVV EIIQATIIRQ NQALRLRARK ECWDRDGKER VTGEEWLVTT VGAYLPAVFE EVLDLVDAVI LTEKTALHLR ARRNFRDFRG VSRRTGEEWL VTVQDTEAHV PDVHEEVLGV VPITTLGPHN YCVILDPVGP DGKNQLGQKR VVKGEKSFFL QPGEQLEQGI QDVYVLSEQQ GLLLRALQPL EEGEDEEKVS HQAGDHWLIR GPLEYVPSAK VEVVEERQAI PLDENEGIYV QDVKTGKVRA VIGSTYMLTQ DEVLWEKELP PGVEELLNKG QDPLADRGEK DTAKSLQPLA PRNKTRVVSY RVPHNAAVQV YDYREKRARV VFGPELVSLG PEEQFTVLSL SAGRPKRPHA RRALCLLLGP DFFTDVITIE TADHARLQLQ LAYNWHFEVN DRKDPQETAK LFSVPDFVGD ACKAIASRVR GAVASVTFDD FHKNSARIIR TAVFGFETSE AKGPDGMALP RPRDQAVFPQ NGLVVSSVDV QSVEPVDQRT RDALQRSVQL AIEITTNSQE AAAKHEAQRL EQEARGRLER QKILDQSEAE KARKELLELE ALSMAVESTG TAKAEAESRA EAARIEGEGS VLQAKLKAQA LAIETEAELQ RVQKVRELEL VYARAQLELE VSKAQQLAEV EVKKFKQMTE AIGPSTIRDL AVAGPEMQVK LLQSLGLKST LITDGSTPIN LFNTAFGLLG MGPEGQPLGR RVASGPSPGE GISPQSAQAP QAPGDNHVVP VLR
This gene encodes the major vault protein which is a lung resistance-related protein. Vaults are multi-subunit structures that may be involved in nucleo-cytoplasmic transport. This protein mediates drug resistance, perhaps via a transport process. It is widely distributed in normal tissues, and overexpressed in multidrug-resistant cancer cells. The protein overexpression is a potentially useful marker of clinical drug resistance. This gene produces two transcripts by using two alternative exon 2 sequences; however, the open reading frames are the same in both transcripts.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Lung resistance-related protein; Major vault protein; MVP; MVP1; Testicular Secretory Protein Li 30; VAULT1; vRNP
Gene Aliases: LRP; MVP; VAULT1
UniProt ID: (Human) Q14764
Entrez Gene ID: (Human) 9961
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support