Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
MYH10 encodes a member of the myosin superfamily. The protein represents a conventional non-muscle myosin; it should not be confused with the unconventional myosin-10 (MYO10). Myosins are actin-dependent motor proteins with diverse functions including regulation of cytokinesis, cell motility, and cell polarity. Mutations in this gene have been associated with May-Hegglin anomaly and developmental defects in brain and heart. Multiple transcript variants encoding different isoforms have been found for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Cellular myosin heavy chain, type B; flectin; MCH-B(B2); Myosin heavy chain 10; myosin heavy chain 10, non-muscle; Myosin heavy chain, non-muscle IIb; myosin heavy chain, nonmuscle type B; myosin heavy chain, nonmuscular, type B; myosin IIB; myosin, heavy polypeptide 10, non-muscle; Myosin-10; NMMHC II-b; NMMHC IIB; NMMHC-B; NMMHC-IIB; Non-muscle myosin heavy chain B; Non-muscle myosin heavy chain IIb; nonmuscle myosin; nonmuscle myosin heavy chain II-B; nonmuscle myosin heavy chain IIB; nonmuscle myosin heavy chain-B; nonmuscle myosin II heavy chain-B; OTTHUMP00000165799
Gene Aliases: 5730504C04Rik; 9330167F11Rik; Fltn; MCH-B; mKIAA3005; MYH10; Myhn-2; Myhn2; NMHC II-B; NMHC-B; NMHCII-B; NMMHC II-b; NMMHC-B; NMMHC-IIB; NMMHCB; SMemb
UniProt ID: (Human) P35580, (Mouse) Q61879, (Rat) Q9JLT0
Entrez Gene ID: (Human) 4628, (Mouse) 77579, (Rat) 79433
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support