Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MSSDSEMAIF GEAAPFLRKS EKERIEAQNK PFDAKTSVFV VDPKESYVKA IVQSREGGKV TAKTEAGATV TVKEDQVFSM NPPKYDKIED MAMMTHLHEP AVLYNLKERY AAWMIYTYSG LFCVTVNPYK WLPVYNPEVV TAYRGKKRQE APPHIFSISD NAYQFMLTDR ENQSILITGE SGAGKTVNTK RVIQYFATIA VTGEKKKEEP ASGKMQGTLE DQIISANPLL EAFGNAKTVR NDNSSRFGKF IRIHFGATGK LASADIETYL LEKSRVTFQL KAERSYHIFY QILSNKKPEL IEMLLITTNP YDFAFVSQGE ITVPSIDDQE ELMATDSAVD ILGFTADEKV AIYKLTGAVM HYGNMKFKQK QREEQAEPDG TEVADKAAYL TSLNSADLLK SLCYPRVKVG NEFVTKGQTV QQVYNAVGAL AKAIYEKMFL WMVTRINQQL DTKQPRQYFI GVLDIAGFEI FDFNSLEQLC INFTNEKLQQ FFNHHMFVLE QEEYKKEGIE WEFIDFGMDL AACIELIEKP MGIFSILEEE CMFPKATDTS FKNKLYEQHL GKSNNFQKPK PAKGKPEAHF SLVHYAGTVD YNIAGWLDKN KDPLNETVVG LYQKSAMKTL AFLFSGAQTA EAEGGGGKKG GKKKGSSFQT VSALFRENLN KLMTNLRSTH PHFVRCIIPN ETKTPGAMEH ELVLHQLRCN GVLEGIRICR KGFPSRILYA DFKQRYKVLN ASAIPEGQFI DSKKASEKLL GSIEIDHTQY KFGHTKVFFK AGLLGTLEEM RDEKLAQLIT RTQAICRGFL MRVEFRKMME RRESIFCIQY NIRAFMNVKH WPWMKLYFKI KPLLKSAETE KEMANMKEEF EKTKEELAKT EAKRKELEEK MVTLMQEKND LQLQVQAEAD ALADAEERCD QLIKTKIQLE AKIKEVTERA EDEEEINAEL TAKKRKLEDE CSELKKDIDD LELTLAKVEK EKHATENKVK NLTEEMAGLD ETIAKLTKEK KALQEAHQQT LDDLQMEEDK VNTLTKAKTK LEQQVDDLEG SLEQEKKLCM DLERAKRKLE GDLKLAQEST MDTENDKQQL NEKLKKKEFE MSNLQGKIED EQALAIQLQK KIKELQARIE ELEEEIEAER ASRAKAEKQR SDLSRELEEI SERLEEAGGA TSAQIEMNKK REAEFQKMRR DLEESTLQHE ATAAALRKKH ADSVAELGEQ IDSLQRVKQK LEKEKSELKM EINDLASNME TVSKAKANFE KMCRTLEDQL SEIKTKEEEQ QRLINELSAQ KARLHTESGE FSRQLDEKDA MVSQLSRGKQ AFTQQIEELK RQLEEETKAK STLAHALQSA RHDCDLLREQ YEEEQEAKAE LQRGMSKANS EVAQWRTKYE TDAIQRTEEL EEAKKKLAQR LQDAEEHVEA VNSKCASLEK TKQRLQNEVE DLMIDVERSN AACIALDKKQ RNFDKVLAEW KQKYEETQAE LEASQKESRS LSTELFKVKN AYEESLDHLE TLKRENKNLQ QEISDLTEQI AEGGKHIHEL EKVKKQLDHE KSELQTSLEE AEASLEHEEG KILRIQLELN QVKSEIDRKI AEKDEELDQL KRNHLRVVES MQSTLDAEIR SRNDALRIKK KMEGDLNEME IQLNHANRQA AEALRNLRNT QGILKDTQLH LDDAIRGQDD LKEQLAMVER RANLMQAEVE ELRASLERTE RGRKMAEQEL LDASERVQLL HTQNTSLINT KKKLETDISQ IQGEMEDIVQ EARNAEEKAK KAITDAAMMA EELKKEQDTS AHLERMKKNM EQTVKDLQLR LDEAEQLALK GGKKQIQKLE ARVRELESEV ESEQKHNVEA VKGLRKHERR VKELTYQTEE DRKNILRLQD LVDKLQTKVK AYKRQAEEAE EQSNVNLAKF RKLQHELEEA KERADIAESQ VNKLRVKSRE VHTKVISEE
Target protein includes a fast myosin isoform expressed in skeletal muscle and has function involved in Muscle contraction.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: MyHC-2b; MyHC-IIb; Myosin heavy chain 2b; Myosin heavy chain 4; Myosin heavy chain IIb; Myosin heavy chain, skeletal muscle, fetal; myosin, heavy chain 4, skeletal muscle; myosin, heavy polypeptide 4, skeletal muscle; Myosin-4
Gene Aliases: MYH2B; MYH4; MyHC-2B; MyHC-IIb
UniProt ID: (Human) Q9Y623
Entrez Gene ID: (Human) 4622
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support