Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Positive Samples: Mouse skeletal muscle
Immunogen sequence: MTDAQMADFG AAAQYLRKSE KERLEAQTRP FDIRTECFVP DDKEEFVKAK ILSREGGKVI AETENGKTVT VKEDQVLQQN PPKFDKIEDM AMLTFLHEPA VLFNLKERYA AWMIYTYSGL FCVTVNPYKW LPVYNAEVVA AYRGKKRSEA PPHIFSISDN AYQYMLTDRE NQSILITGES GAGKTVNTKR VIQYFASIAA IGDRGKKDNA NANKGTLEDQ IIQANPALEA FGNAKTVRND NSSRFGKFIR
Cardiac muscle myosin is a hexamer consisting of two heavy chain subunits, two light chain subunits, and two regulatory subunits. This gene encodes the alpha heavy chain subunit of cardiac myosin. The gene is located approximately 4kb downstream of the gene encoding the beta heavy chain subunit of cardiac myosin. Mutations in this gene cause familial hypertrophic cardiomyopathy and atrial septal defect 3.
⚠WARNING: This product can expose you to chemicals including mercury, which is known to the State of California to cause birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: alpha cardiac MHC; alpha myosin; cardiac myosin heavy chain alpha isoform; cardiomyopathy, hypertrophic 1; MyHC-alpha; myosin 6; Myosin heavy chain 6; myosin heavy chain polypeptide 6 cardiac muscle adult; Myosin heavy chain, cardiac muscle alpha isoform; Myosin heavy chain, cardiac muscle alpha isoform (MyHC-alpha); myosin heavy chain, cardiac muscle, adult; myosin heavy chain, polypeptide 6, cardiac muscle, alpha; myosin, heavy polypeptide 6, cardiac muscle, alpha; Myosin-6
Gene Aliases: A830009F23Rik; AA517445; alpha-MHC; Myh6; Myhc-a; Myhca
UniProt ID: (Mouse) Q02566
Entrez Gene ID: (Mouse) 17888, (Rat) 29556
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support