Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: DEMIKEAPGP INFTVFLTMF GEKLKGADPE ETILNAFKVF DPEGKGVLKA DYVREMLTTQ AERFSKEEVD QMFAAFPPDV TGNLDYKNLV HIITHGEEKD
Thus gene encodes the regulatory light chain associated with cardiac myosin beta heavy chain. Ca+ triggers the phosphorylation of regulatory light chain that in turn triggers contraction. Mutations in this gene are associated with mid-left ventricular chamber type hypertrophic cardiomyopathy.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: cardiac myosin light chain-2; DKFZp779C0562; MLC-2; MLC-2s/v; Myosin light chain 2, slow skeletal/ventricular muscle isoform; myosin light chain 2v; myosin light chain phosphatase; myosin light chain, phosphorylatable, cardiac ventricles; Myosin regulatory light chain 2, ventricular/cardiac muscle isoform; myosin, light polypeptide 2, regulatory, cardiac, slow; ventricular myosin regulatory light chain
Gene Aliases: MLC-2; MLC-2v; Mlc2; Mlc2v; Myl2; Mylpc
UniProt ID: (Rat) P08733, (Mouse) P51667
Entrez Gene ID: (Rat) 363925, (Mouse) 17906
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support