Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Immunogen sequence: MELLSPPLR DVDLTAPDGS LCSFATTDDF YDDPCFDSPD LRFFEDLDPR LMHVGALLKP EEHSHFPAAV HPAPGAREDE HVRAPSGHHQ AGRCLLWACK ACKRKTTNAD RRKAATMRER RRLSKVNEAF ETLKRCTSSN PNQRLPKVEI LRNAIRYIEG LQALLRDQDA APPGAAAAFY APGPLPPGRG GEHYSGDSDA SSPRSNCSDG MMDYSGPPSG ARRRNCYEGA YYNEAPSEPR PGKSAAVSSL DCLSSIVERI STESPAAPAL LLADVPSESP PRRQEAAAPS EGESSGDPTQ SPDAAPQCPA GANPNPIYQV L (1-320 aa encoded by BC064493)
The Myogenic determination gene (MyoD) was first identified by the virtue of its ability to convert embryonic mouse fibroblast cells to muscle cells. It was subsequently shown that forced expression of MyoD (human homolog is myf 3) gene in a wide variety of normal and neoplastic cells could either convert the cells to muscle cells or activate a set of the otherwise transcriptionally inactive muscle-specific genes in these cells. The regulatory domain of the MyoD gene product lies within a 70 amino acid region and comprises a basic DNA binding motif and a helix-loop-helix (HLH) dimerization motif. Subsequent studies identified three other genes whose products shared sequence homology for the basic HLH domain of MyoD. These are; myf5, myogenin (human homolog is myf4) and myf6 (also known as MRF4 and herculin).
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: bHLHc1; Class C basic helix-loop-helix protein 1; eukaryotic myogenic factor MYF-3; Myf-3; Myoblast determination protein 1; Myogenic factor 3; myogenic regulatory factor
Gene Aliases: AI503393; BHLHC1; MUF3; MYF-3; MYF3; MYOD; Myod-1; MYOD1; PUM
UniProt ID: (Human) P15172, (Pig) P49811, (Mouse) P10085, (Rat) Q02346
Entrez Gene ID: (Human) 4654, (Pig) 407604, (Mouse) 17927, (Rat) 337868
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support