Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
NeoBiotechnologies
FIGURE: 1 / 3
Current literature indicates that mammaglobin, as measured by the ELISA, holds significant promise for breast cancer screening with the realistic potential to impact management of this disease.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: mammaglobin 1; Mammaglobin-1; Mammaglobin-A; Secretoglobin family 2A member 2; secretoglobin, family 2A, member 2
Gene Aliases: MGB1; SCGB2A2; UGB2
UniProt ID: (Human) Q13296
Entrez Gene ID: (Human) 4250
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support