Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
FIGURE: 1 / 2
Specificity: This antibody binds the extracellular domain III of the human EGFR and does not cross react with murine EGFR. It was also reported that residues S460/G461 in EGFR domain III are essential components of the epitope. This antibody binds at a site distinct from the EGF binding site, preventing conformational re-arrangement required for dimerization, a process essential for high-affinitiy ligand binding. This antibody can only recognize the active form of EGFR. The antibody binds to both glyco- and aglyco receptor forms.
Matuzumab is a humanized monoclonal antibody used in cancer treatment. It has a high affinity for EGFR, frequently associated with the growth of blood vessels in malignancy, facilitating tumor growth and survival.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: EMD-72000; H425
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support