Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 2
Positive test controls include: HeLa, HCT116, HepG2, NIH/3T3, Mouse lung, Mouse brain, Rat lung, Rat brain. The target is usually found in the following locations: Nucleus.
Immunogen sequence: VSGPPRKPPG TVAGTARGPE GGSTAQVPAP TASPPPEGPV LTFQSEKMKG MKELLVATKI NSSAIKLQLT AQSQVQMKKQ KVSTPSDYTL SFLKRQRKGL
Potassium channels are a group of ubiquitously expressed proteins that serve numerous functions in excitable and non-excitable cells. One class of integral membrane potassium channels is the large conductance, calcium-activated potassium channel (Maxi K+). Maxi K+ differs from most other potassium channels in that its activation is controlled by both increases in intracellular calcium and by membrane depolarization. Maxi K+ dual activation is possible because of its structure. The core of the channel, which is similar to other potassium channels, is a Maxi K+ alpha homotetramer that contains both a voltage sensor and an intracellular calcium binding domain. In vascular smooth muscle, an auxiliary beta-subunit is found in a 1:1 stoichiometry. The beta-subunit exhibits its effect on the Maxi K+ channel by effectively decreasing by 5- to 10- fold the concentration of calcium required to keep the pore open.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Menin; multiple endocrine neoplasia 1; multiple endocrine neoplasia I
Gene Aliases: AW045611; MEAI; MEN1; menin; SCG2
UniProt ID: (Human) O00255, (Mouse) O88559, (Rat) Q9WVR8
Entrez Gene ID: (Human) 4221, (Mouse) 17283, (Rat) 29417
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support