Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Metallothionein (MT) is a sulfhydryl- and cysteine-rich protein found in micro-organisms, plants and all invertebrate and vertebrate animals. Metallothioneins are a group of ubiquitous low-molecular-weight proteins that have functional roles in cell growth, repair and differentiation. Metallothionein are implicated primarily in metal ion detoxification as they are essential for the protection of cells against the toxicity of cadmium, mercury and copper. Metallothioneins are known to be broadly expressed in heart, liver, kidney, breast and testis tissue. Metallothionein 3, also known as MT-3 or GIFB (growth inhibitory factor), is a 68 amino acid protein that belongs to the type 1 family and Metallothionein superfamily. While highly expressed in astrocytes of the normal human brain, Metallothionein 3 expression is reduced in the brains of patients with Alzheimer disease.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: GIF; GIFB; Growth inhibitory factor; metallothionein 3 (growth inhibitory factor (neurotrophic)); metallothionein III; Metallothionein-3; Metallothionein-III; MT-3; MT-III
Gene Aliases: GIF; GIFB; GRIF; Mt-3; MT3; ZnMT3
UniProt ID: (Human) P25713, (Rat) P37361, (Mouse) P28184
Entrez Gene ID: (Human) 4504, (Rat) 117038, (Mouse) 17751
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support