Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 1
Immunogen sequence: SYVSDQNGGV PDPNPPPPQT HRRLRRSHSG PAGSFNKPAI RISNCCIHRA SPNSDEDKPE VPPRVPIPPR PVKPDYRRWS AEVTSSTYSD EDRPPKVPPR EPLSPSNSRT PSPKSLPSYL NGVMPPTQSF APDPKYVSSK ALQRQNSEGS ASKVPCILPI IENGKKVSST HYYLLPERPP YLDKYEKFFR EAEETNGGAQ IQPLPADCGI SSATEKPDSK TKMDLGGHVK RKHLSYVVSP; Positive Samples: Rat lung; Cellular Location: Cell membrane, Cytoplasm, Cytoplasmic side, Nucleus, Peripheral membrane protein
ERRFI1 is a cytoplasmic protein whose expression is upregulated with cell growth (Wick et al., 1995). It shares significant homology with the protein product of rat gene-33, which is induced during cell stress and mediates cell signaling (Makkinje et al., 2000).
⚠WARNING: This product can expose you to chemicals including mercury, which is known to the State of California to cause birth defects or other reproductive harm. For more information go to www.P65Warnings.ca.gov.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ERBB receptor feedback inhibitor 1; Gene 33 polypeptide; MIG-6; Mitogen-inducible gene 6 protein homolog; RALT; Receptor-associated late transducer
Gene Aliases: 33; Errfi1; gene 33; Mig6; Ralt; RGD1307599; Xxx
UniProt ID: (Rat) P05432
Entrez Gene ID: (Rat) 313729
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support