Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 4
Immunogen sequence: KTRAELKTAM STPHVAEPAE NEQDEQDENG AEASADLRAD AMAKDRSEEE RTTEAEKNER VQKHLKALTS ELANARDESK KTANDMIHAE NMRLGRDKYK; Positive Samples: HeLa, Jurkat, HepG2, DU145, HT1080, NIH/3T3, Mouse thymus, Mouse lung, Rat lung; Cellular Location: Apical cell membrane, Cell membrane, Cell projection, Cytoplasm, Cytoplasmic side, Peripheral membrane protein, cytoskeleton, microvillus membrane
Moesin, a member of the talin-4. 1 superfamily, is a linking protein of the submembraneous actin cytoskeleton. It is expressed in macrophages, lymphocytes, fibroblastic, endothelial, epithelial, and neuronal cell lines but not in blood cells. It is involved in cell adhesion, migration, and organization of cell surface structures.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: epididymis luminal protein 70; Membrane-organizing extension spike protein; Moesin
Gene Aliases: C78546; HEL70; MSN
UniProt ID: (Human) P26038, (Rat) O35763, (Mouse) P26041
Entrez Gene ID: (Human) 4478, (Rat) 81521, (Mouse) 17698
Molecular Function:
actin or actin-binding cytoskeletal protein
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support