Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AbboMax
FIGURE: 1 / 1
Positive control: 3T3 Nuclear Extract
Cellular location: Nucleus.
Histones are basic nuclear proteins that together with DNA make up the nucleosome structure in eukaryotes. Nucleosomes consist of approximately 146 bp of DNA wrapped around a histone octamer composed of pairs of each of the four core histones (H2A, H2B, H3, and H4). The methylation of specific residues in the histone tails is a central modification for regulatingepigenetic transitions in chromatin. Whereas methylation of histone H3 on lysine 4, 36, and 79 has been linked with gene activation, methylation of H3 on lysines 9, 27 and histone H4 on lysine 20 is associated with heterochromatin and some repressed genes with euchromatin. Modified lysine residues can exist in a mono-, di-, or tri-methylated state, while the arginine residues can be mono- or di-methylated. Histone H3 Lys4 trimethylation (H3-K4me3) is a conserved mark of actively transcribed chromatin. This antibody is specific for histone H3 tri-methylated at K27. The sequence is found in all mammals and a wide range of species, including D. melanogaster, Arabidopsis, Chicken and Xenopus. The antibody will react with any of the above species where the trimethylation modification is present.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: H3 histone family, member J; histone 1, H3j; Histone H3.1; Histone H3/a; Histone H3/b; Histone H3/c; Histone H3/d; Histone H3/f; Histone H3/h; Histone H3/i; Histone H3/j; Histone H3/k; Histone H3/l
Gene Aliases: H3/j; H3C1; H3C10; H3C11; H3C12; H3C2; H3C3; H3C4; H3C6; H3C7; H3C8; H3FA; H3FB; H3FC HIST1H3C; H3FD; H3FF; H3FH; H3FI; H3FJ; H3FK; H3FL; HIST1H3A; HIST1H3B; HIST1H3D; HIST1H3E; HIST1H3F; HIST1H3G; HIST1H3H; HIST1H3I; HIST1H3J
UniProt ID: (Human) P68431
Entrez Gene ID: (Human) 8356
Molecular Function:
histone
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support