Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
Immunogen sequence: LVLANPESAT TFKVLAAKVL NLVLPNLSLG PIDSSVLSRN KTEVDIYNSD PLICRAGLKV CFGIQLLNAV SRVERALPKL TVPFLLLQGS ADRLCDSKGA YLLMELAKSQ DKTLKIYEGA YHVLHKELPE VTNSVFHE
Antigen sequence identity: Mouse 86%; Rat 86%
Monoglyceride lipase functions together with hormone-sensitive lipase to hydrolyze intracellular triglyceride stores in adipocytes and other cells to fatty acids and glycerol. MGLL may also complement lipoprotein lipase in completing hydrolysis of monoglycerides resulting from degradation of lipoprotein triglycerides.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: HU-K5; Lysophospholipase homolog; Lysophospholipase-like; MAGL; MGL; Monoacylglycerol lipase; Monoglyceride lipase
Gene Aliases: HU-K5; HUK5; MAGL; MGL; MGLL
UniProt ID: (Human) Q99685
Entrez Gene ID: (Human) 11343
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support