Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Detects myozap (myocardium zonula adherens protein), a major 54 kD polypeptide present in myocardial junctions (composite junctions in the intercalated disks), also localized in adherens type junctions of the entire cardiovascular system and in endothelia of the blood and lymph vascular systems.Reactivity on cultured cell lines: HL-1, HUVEC.Positive control: myocard.
This gene (Gcom1) is part of a complex transcript unit that includes the gene for glutamate receptor, ionotropic, N-methyl D-aspartate-like 1A (GRINL1A). Transcription of this gene occurs at an upstream promoter, with two different groups of alternatively spliced variants: Gup for GRINL1A upstream transcripts and Gcom for GRINL1A combined transcripts. The GRINL1A gene uses a downstream promoter for transcription and also has multiple alternatively spliced variants.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: DNA-directed RNA polymerase II subunit GRINL1A; DNA-directed RNA polymerase II subunit GRINL1A, isoforms 4/5; DNA-directed RNA polymerase II subunit M; DNA-directed RNA polymerase II subunit M, isoforms 4/5; gcom; Gcom2; Gdown1; glutamate receptor, ionotropic, N-methyl D-aspartate-like 1A; Glutamate receptor-like protein 1A; GRINL1A downstream protein 1; GRINL1A downstream protein Gdown4; GRINL1A upstream protein; Gup; Myocardial zonula adherens protein; MYZAP-POLR2M; polymerase (RNA) II (DNA directed) polypeptide M; polymerase (RNA) II subunit M; protein GRINL1A; protein GRINL1A, isoforms 4/5
Gene Aliases: AA407243; AA408799; AI303746; AW553238; D9Wsu138e; GCOM1; Gdown; Gdown1; Glurr; GRINL1A; mKIAA4185; MYOZAP; MYZAP; POLR2M
UniProt ID: (Bovine) Q17QE3, (Human) P0CAP2, (Mouse) Q6P6I6, (Rat) Q91XQ4
Entrez Gene ID: (Bovine) 100125877, (Human) 81488, (Pig) 102160778, (Mouse) 28015, (Rat) 192147
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support