Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
The antibody detects a 215 kDa* protein corresponding to the apparent molecular mass of MYH4 on SDS-PAGE immunoblots of mouse extraocular muscle and mouse C2C12 cells. The antibody may also detect other skeletal MYHs, such as MYH1 and MYH2, which have significant homology to the C-terminal region of MYH4.
Target protein includes a fast myosin isoform expressed in skeletal muscle and has function involved in Muscle contraction.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 2B myosin heavy chain; myh4; MyHC-2b; MyHC-IIb; Myosin heavy chain 2b; Myosin heavy chain 4; Myosin heavy chain IIb; myosin heavy chain type IIb; Myosin heavy chain, skeletal muscle, fetal; myosin, heavy chain 4, skeletal muscle; myosin, heavy polypeptide 4, skeletal muscle; Myosin-4
Gene Aliases: AI506973; MHC2B; Minimsc; Minmus; MM; MYH-2B; MYH2B; MYH4; MyHC-2B; MyHC-IIb; Myhsf
UniProt ID: (Human) Q9Y623, (Mouse) Q5SX39, (Rat) Q29RW1
Entrez Gene ID: (Human) 4622, (Mouse) 17884, (Rat) 360543
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support