Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
MA5-31804 has been tested in indirect ELISA.
The protein encoded by this gene is a member of the bone morphogenetic protein family and the TGF-beta superfamily. This group of proteins is characterized by a polybasic proteolytic processing site which is cleaved to produce a mature protein containing seven conserved cysteine residues. The members of this family are regulators of cell growth and differentiation in both embryonic and adult tissues. This gene is thought to encode a secreted protein which negatively regulates skeletal muscle growth.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: GDF-8; Growth/differentiation factor 8; H-Myo-Propeptide; H-Myostatin; Myostatin
Gene Aliases: GDF8; MSLHP; MSTN
UniProt ID: (Human) O14793
Entrez Gene ID: (Human) 2660
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support