Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 3
Nociceptin is the ligand of the opioid receptor-like receptor (OPRL1). It may act as a transmitter in the brain by modulating nociceptive and locomotor behavior. May be involved in neuronal differentiation and development.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: N23K/N27K; neuropeptide nociceptin 1; nociceptin; nocistatin; OFQ Orphanin FG; orphanin FQ; pre-pro-N/OFQ; Prepronociceptin; Prepronociceptin (neuropeptide nociceptin) (N23K); pronociceptin; proorphanin
Gene Aliases: N/OFQ; N23K; NOP; Npnc1; OFQ; OFQ/N; PNOC; ppN/OFQ; PPNOC
UniProt ID: (Human) Q13519, (Mouse) Q64387, (Rat) Q62923
Entrez Gene ID: (Human) 5368, (Mouse) 18155, (Rat) 25516
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support